Learn More
Simport™ Scientific Unisette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. 23045568 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-568 Blue
23-045-561 Lilac
23-045-562 Orange
23-045-563 Aqua
23-045-564 White
23-045-565 Pink
23-045-566 Green
23-045-567 Yellow
23-045-569 Peach
23-045-570 Tan
23-045-571 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 23-045-568 Supplier Simport™ Scientific Supplier No. M5266T
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Simport™ Unisette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks is offered with a 45° anterior writing surface. Configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
  • Keeps specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents.
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage.
  • One-piece construction with snap-latch and hinge-lock design allows for simple one-hand operation and prevents early separation of the base and lid.
  • Can be opened and closed as often as necessary and they re-lock securely without the danger of specimen loss.
  • Convenient Quickload™ stacks are suitable for automated printers.
Specifications
Writing Face Angle 45°
Quantity 1000/Cs.
Color Blue
Material Acetal
Product Type Cassette