Learn More
Simport™ Scientific Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. 22038072 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-072 Peach
22-038-067 White
22-038-068 Pink
22-038-069 Green
22-038-070 Yellow
22-038-071 Blue
22-038-073 Tan
22-038-074 Gray
22-038-075 Lilac
22-038-076 Orange
22-038-077 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-038-072 Supplier Simport™ Scientific Supplier No. M5057T
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Secure specimens during processing with Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
  • In Quickload™ stacks
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 25 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 1000/Pk.
Color Peach
Material Acetal