Learn More
Simport™ Scientific Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. 23045558 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-558 Peach
23-045-551 Orange
23-045-552 Aqua
23-045-553 White
23-045-554 Pink
23-045-555 Green
23-045-556 Yellow
23-045-557 Blue
23-045-559 Tan
23-045-560 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 23-045-558 Supplier Simport™ Scientific Supplier No. M5257T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks is offered with a 45° anterior writing surface. Configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
  • Keeps specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents.
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage.
  • One-piece construction with snap-latch and hinge-lock design allows for simple one-hand operation and prevents early separation of the base and lid.
  • Can be opened and closed as often as necessary and they re-lock securely without the danger of specimen loss.
  • Convenient Quickload™ stacks are suitable for automated printers.
Specifications
Writing Face Angle 45°
Quantity 1000/Cs.
Color Peach
Material Acetal
Product Type Cassette