Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL360 UltraLite™ PF Zero Unisex Lab Coats
SDP

Catalog No. 22412387
Change view
Click to view available options
Color:
Black
Ceil Blue
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
36 product options available for selection
Product selection table with 36 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22-412-387 Ceil Blue X-Small
22-412-378 White X-Small
22-412-379 White Small
22-412-380 White Medium
22-412-381 White Large
22-412-382 White X-Large
22-412-383 White 2X-Large
22-412-384 White 3X-Large
22-412-385 White 4X-Large
22-412-386 White 5X-Large
22-412-388 Ceil Blue Small
22-412-389 Ceil Blue Medium
22-412-390 Ceil Blue Large
22-412-391 Ceil Blue X-Large
22-412-392 Ceil Blue 2X-Large
22-412-393 Ceil Blue 3X-Large
22-412-394 Ceil Blue 4X-Large
22-412-395 Ceil Blue 5X-Large
22-412-396 Royal Blue X-Small
22-412-397 Royal Blue Small
22-412-398 Royal Blue Medium
22-412-399 Royal Blue Large
22-412-400 Royal Blue X-Large
22-412-401 Royal Blue 2X-Large
22-412-402 Royal Blue 3X-Large
22-412-403 Royal Blue 4X-Large
22-412-404 Royal Blue 5X-Large
22-412-405 Black X-Small
22-412-406 Black Small
22-412-407 Black Medium
22-412-408 Black Large
22-412-409 Black X-Large
22-412-410 Black 2X-Large
22-412-411 Black 3X-Large
22-412-412 Black 4X-Large
22-412-413 Black 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
36 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 22-412-387 missing translation for 'mfr' DenLine™ Uniforms missing translation for 'supplierNo' DL360CBXS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Breathable lab coats provide AAMI PB70 Level 2 fluid resistance and feature heavy duty stainless steel snap closures and hip side vents.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Dental
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • Complies with OSHA 29 CFR 1910
  • Provides 360 degree front and back, plus shoulder protection
  • AAMI PB70 Level 2
  • Overall Air Permeability 35 CFM (ASTM D737)
  • Stain resistant fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double stitched
  • Stainless snap closures
  • Cuffs for overgloving
  • Bleach safe all colors
  • Hang loop, outside back of collar
  • Side vents for pant pocket access
  • Autoclavable fabric
  • 225 commercial wash tested

Compliance

DL360 UltraLite PF ZERO is Environmentally Friendly, PFOS and PFOA Free. PF ZERO Fabric is treated with non-fluorinated aqueous repellent chemistry where PFAS substances are not intentionally added. Based on PFAS Test Method Minimum Detection Limit.

Specifications

Product Type General Purpose Lab Coat
Size X-Small
Gender Unisex
Material Polyester, Carbon
Color Ceil Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Static, Stains
No. of Pockets 3
Collar Type Traditional
Latex Free Yes
Size (English) Chest 30 to 32 in.
Size (English) Hip 33 to 35 in.
Size (Metric) Chest 76 to 81 cm
Size (Metric) Hip 84 to 89 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanrooms
Includes Hip Side Vents
Length (English) 41 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 35 in.
Length (Metric) 104.1 cm
Length (Metric) Cuff 10.1 cm
Weight (English) 0.06 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 35 CFM (ASTM D737)
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.