Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL470BJ Unisex Long Length White Lab Coats, Cost Effective
SDP

Catalog No. 22412415
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-412-415 Small
22-412-414 X-Small
22-412-416 Medium
22-412-417 Large
22-412-418 X-Large
22-412-419 2X-Large
22-412-420 3X-Large
22-412-421 4X-Large
22-412-422 5X-Large
22-412-423 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 22-412-415 Supplier DenLine™ Uniforms Supplier No. DL470BJWHS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cost effective long length lab coats provide AAMI PB70 Level 2 fluid resistance and feature heavy duty stainless steel snap closures.

  • Primary Applications:
    • Anatomic pathology
    • Clinical pathology and laboratory
    • Phlebotomy
    • Specimen and blood collection
    • Manufacturing
    • Animal research
    • Cleanroom ISO 5, Class 100
  • Highest barrier protection against strikethrough from incidental splash of blood
  • Hydrostatic Pressure 32.5 CM (AATCC 127) and Impact Penetration 1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles
  • Complies with OSHA 29 CFR 1910
  • 360 Degree protection front as well as shoulders and back
  • Fabric is fluid resistant, AAMI PB70 Level 2 Barrier
  • High air permeability (35 CFM) is used to make the back of coat
  • Overall air permeability of coat front and back is 18 CFM (ASTM D737)
  • Stain resistant fabric and cuffs (blood, bleach and all aqueous reagents, liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double-stitched, bar tacked
  • Stainless snap closures
  • Durable cuffs for overgloving, bleach safe all colors
  • Hang loop, outside back of collar
  • Side vents for pant pocket access

Specifications

Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Polyester, Carbon
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Stains, Static
No. of Pockets 3
Collar Type Traditional
Latex Free Yes
Size (English) Chest 34 to 36 in.
Size (English) Hip 36 to 37 in.
Size (Metric) Chest 86 to 91 cm
Size (Metric) Hip 91 to 94 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanrooms
Length (English) 41 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 36.5 in.
Length (Metric) 104.1 cm
Length (Metric) Cuff 10.1 cm
Weight (English) 0.7 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 18 CFM (ASTM D737)
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.