Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Unisex Cotton Lab Coats

Catalog No. 18111227
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-227 X-Large
18-111-222 2X-Small
18-111-223 X-Small
18-111-224 Small
18-111-225 Medium
18-111-226 Large
18-111-228 2X-Large
18-111-229 3X-Large
18-111-230 4X-Large
18-111-231 5X-Large
18-111-232 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 18-111-227 Supplier CritiCore Protective Wear Supplier No. 12L24884100C3PXLWHT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unisex Lab Coats are made of breathable, comfortable, 100% cotton fabric and feature a quick-release five-snap front and knit cuffs.

  • Quick-release 5-snap front
  • Knit cuffs
  • Breast pocket with pencil slot
  • 2 oversized waist pockets
  • 100% cotton

Specifications

Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material Cotton
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Lapel
For Use With (Application) General Laboratory, Medical, Healthcare
Length (English) 39 in.
Length (English) Sleeve 25 in.
Length (Metric) 99.06 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.