Learn More
Synbiosis™ UNISTAT™ Bioassay Analysis Software
GREENER_CHOICESDP
Catalog No. 01257321 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
ProtoCOL 3 Instruments
Catalog No. For Use With (Equipment)
01-257-321 ProtoCOL 3 Instruments
1 options
Catalog No. 01-257-321 Supplier Synbiosis™ Supplier No. SYNBBIOSTAT
Add to Cart
Add to Cart
UNISTAT™ Bioassay Analysis Software has been integrated for use with ProtoCOL 3 colony counting and inhibition zone measuring system. The statistical module allows scientists a quick, easy method of analyzing their ProtoCOL 3 data to generate accurate antibiotic and vaccine potency results.
UNISTAT™ Bioassay Analysis Software has been integrated for use with ProtoCOL 3, allowing microbiologists to rapidly export and analyze inhibition zone measurement and colony count data.
  • Accurate reports are produced quickly and easily, making this software ideal for users whose primary training is not in statistics
  • Analyze data from bioassays in a single platform, including:
    • Parallel Line Assay: Completely Randomized Design, Randomized Block Design, Latin Squares Design, Crossover Design
    • Slope Ratio Assay: Blanks, Plate Effects
    • Quantal Response Assay: Logit, Probit, Gompit, Loglog link functions Four-Parameter Logistic Model: European Pharmacopoeia (EP), Full Model (USP), Reduced Model (USP)
    • Combination of Assays: EP, USP
    • Cylinder-Plate (5+1) Assay
  • Regular upgrades and long term, expert support available
  • Meets the requirements of common pharmacopeia, following the methods required by EP and USP
Specifications
For Use With (Application) Statistical package for determining vaccine and antibiotic potency
Product Type Statistics Software Package
For Use With (Equipment) ProtoCOL 3 Instruments