Learn More
United Scientific™ UniStore™ Jerricans
SDP
Catalog No. 01676792 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01-676-792 20 L
01-676-791 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-676-792 Supplier United Scientific™ Supplier No. 35020
Add to Cart
Add to Cart
Jerrican for storage and transport of liquids.
UniStore™ jerricans are specialized containers designed for the safe storage, transport, and dispensing of liquid chemicals, reagents, and hazardous materials. These jerricans are constructed from high-density polyethylene (HDPE) and have excellent chemical resistance, impact strength, and long-term durability. The jerricans feature a comfortable handle, angled spout for accurate pouring, leakproof closure, stable base, and precision laser-printed graduations. Jerricans are available in 10 L and 20 L sizes.
  • US FDA Compliant, USP Class VI materials
  • TSE/BSE compliant, BPA-free
  • Manufactured in a cleanroom facility
  • Manufacturing facility compliant to ISO 10993 standards
  • Leakproof polypropylene closure design with silicone gasket
  • Laser-printed durable graduations with lot numbers for traceability
  • Excellent chemical resistance
  • Angled spout for accurate pouring
  • Stable base
  • Robust shoulder handle for ease of carrying
  • Semi-buttress thread design on bottle/closure prevents over-tightening and prevents leaks
  • Rectangular design for efficient use of storage space
  • Jerricans are available in 10 Liter and 20 Liter sizes
  • Jerrican containers are made of HDPE with PP closures
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 20 L
Color White
Material HDPE
Mouth Style Narrow
Product Type Jerry Can
Sterility Non-sterile