Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore Wide Mouth Square Bottles PPCO
SDPSCI_ED

Catalog No. S01879 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01879 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01879 Supplier United Scientific™ Supplier No. 33607
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These wide mouth square bottles are are made of polypropylene copolymer PPCO bottles and are autoclavable.

UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The square design allows the bottles to nest together for more efficient storage, packing and shipping. The polypropylene copolymer (PPCO) bottles are autoclavable at 121°C. These bottles are leakproof and have a polypropylene cap.

  • Square design for compact storage
  • Better impact strength and durability than standard polypropylene (PP)
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces
  • FDA-21CFR compliant resin
  • Autoclavable at 121°C
  • Leakproof closure
  • Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL

Specifications

Capacity (Metric) 250 mL
Color Translucent White
Material Plastic
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Autoclavable Autoclavable
Capacity (English) 8 oz.
Product Type Reagent Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.