Learn More
United Scientific Supplies United Scientific™ Advanced Electricity and Magnetism Kit

Shop All United Scientific Supplies, Inc Products


Description
- 25 activities addressing 7 key topics
- Appropriate for Elementary to High School
- Aligned with NGSS Standards
- Includes detailed Activity Guide
Specifications
Specifications
| Grade Level | Intermediate - Middle School, High School, Higher Ed |
| Class Size | 1 Lab Group |
| Product Type | Electricity and Magnetism Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.