Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Ball and Ring Apparatus
SDPSCI_ED

Catalog No. S13050 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13050 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S13050 Supplier United Scientific Supplies Supplier No. BRA002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Activity guide

Made of brass completely with brown lacquer finish wood handle.

  • Designed to demonstrate thermal expansion.
  • Includes a brass ball and a brass ring each mounted on a handle.
  • The ball fits easily through the ring at room temperature. When the ball is heated over a flame, it expands and no longer fits in the ring.
  • Includes activity guide.

Specifications

Grade 6 to 12
Format Kit
Product Type Ball and Ring Apparatus
Includes Activity guide
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.