Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Ballistics Car with Remote Trigger
SDPSCI_ED

Catalog No. S13064 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13064 5 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13064 Supplier United Scientific Supplies Supplier No. BLCAR2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Conical rubber catcher, two 0.75 in. steel balls, operating instructions

Requires

One 9V battery (not included)

Demonstrates independence of vertical and horizontal motions.

  • Consists of four-wheeled cart carrying vertical tube with spring-loaded piston inside
  • Tube is loaded with ball and cart is set in motion
  • During motion, piston is released remotely by wireless signal, projecting ball upwards out of tube
  • Both ball and cart share same horizontal velocity, which is unaffected by gravity, so that when ball falls back down, it lands back in tube and does not get left behind by cart

Specifications

Grade 5 to 15
Format Kit
Product Type Ballistics Car with Remote Trigger
Includes Conical rubber catcher, two 0.75 in. steel balls, operating instructions
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.