Learn More
United Scientific Supplies United Scientific™ Barnes Dropping Bottle
SDPSCI_ED
Catalog No. S001385 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
30 mL
Catalog No. Capacity (Metric)
S001385 30 mL
1 options
Prop 65
Catalog No. S001385 Supplier United Scientific Supplies Supplier No. BOT030PK/36
Add to Cart
Add to Cart
Prop 65
Barnes dropping bottle includes a glass pipet and rubber bulb stopper for controlled drop delivery.
Barnes Dropping Bottle is designed for laboratory and classroom use. The square-shaped clear flint glass bottle includes a glass pipette and rubber bulb stopper with a 30 mL capacity.
  • Square-shaped clear flint glass bottle
  • Includes glass pipette and rubber bulb stopper
  • 30 mL capacity
  • Replacement pipettes, bulbs, and stoppers available
Specifications
Capacity (Metric) 30 mL
Color Clear
Material Flint Glass
Sterility Non-sterile
Closure Included Yes
Graduated No
Chemical Label Unlabeled
Autoclavable Not Autoclavable
Shape Square
Disposable Reusable
Product Type Dropping Bottle