Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Burette Clamp
SDPSCI_ED

Catalog No. S12872 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S12872 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S12872 Supplier United Scientific™ Supplier No. COBR3U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Burette Clamp for holding small objects.

  • Burette Clamp with boss head.
  • Uncoated jaws.
  • Rotatable jaws.
  • Can be locked at any angle.
  • Clamp attaches to rods up to 13 mm (0.5 in.) outer dia.

Specifications

Type Burette Clamp
Material Alloy Steel
Holds Rods up to 13 mm (0.5 in.) outer dia.
For Use With (Application) Holding small objects
Color Silver
Jaw Type 2-Prong
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.