Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Cavendish Gravitational Balance
SDPSCI_ED

Catalog No. S07390 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07390 5 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07390 Supplier United Scientific™ Supplier No. CGB001
Add to Cart
Add to Cart

This Cavendish Gravitational Balance verifies Newton's Law of Gravity and measures the universal gravitational constant, G.

  • Small scale version.
  • Measures tiny value of G.
  • For advanced Physics labs.

Specifications

Grade 5 to Undergraduate
Format Kit
Product Type Cavendish Gravitational Balance
Languages English
For Use With (Application) Verifies Newton's Law of Gravity and measures the universal gravitational constant, G
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.