Learn More
United Scientific Supplies Clear Base Vacuum Desiccators
SDPSCI_ED
Catalog No. p-5186936 Shop All United Scientific Supplies, Inc Products
Catalog No. S13075 Supplier United Scientific Supplies Supplier No. 55214
Only null left
Add to Cart
Add to Cart
Made of polypropylene (PP) and polycarbonate (PC).
  • Can hold vacuum up to 740mmHg for 24 hours without any greasing
  • Autoclavable at 120°C for 15 minutes
  • Top dome, molded in rigid and transparent PC, gives crystal-clear view of sample
  • Knurled knob on top provides easy handling of dome
  • Transparent PC bottom provides increased viewing clarity
  • Internal groove on flange holds silicon rubber O-ring
  • PP stopcock has self-lubricating PTFE plug
  • Three-way plug opens to create vacuum, closes to hold vacuum, and opens again to release vacuum
Specifications
Product Type Desiccator
Diameter (English) Inner Flange: 5.9 in.
Diameter (Metric) Outer 152.4 mm