Learn More
United Scientific Supplies United Scientific™ Economy Double Burette Clamp with Stand
SDP
Catalog No. 01672027 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Type:
Double Buret Clamp
Catalog No. Type
01-672-027 Double Buret Clamp
1 options
Prop 65
Catalog No. 01-672-027 Supplier United Scientific Supplies Supplier No. UNBURDDSET
Add to Cart
Add to Cart
Prop 65
Double clamp with stand holds burettes up to 30 mm on a 36 in. support rod with a heavy steel base.
  • Stamped steel bases with black enamel finish measures 6 x 11 in.
  • 0.50 in. diameter stainless steel support rod measuring 36 in. long
  • Rubber sleeves
  • For use with: Burettes
Specifications
Material Die-cast Aluminum
Type Double Buret Clamp