Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Energy Conversion Kit
SDPSCI_ED

Catalog No. S05818 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 to 2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05818 1 to 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05818 Supplier United Scientific™ Supplier No. ECK001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Energy Conversion Kit is an economical seven-piece kit demonstrates the conversion among forms of energy.

Four devices for generating electrical energy from:

  • Mechanical motion—hand generator
  • Chemical energy—batteries
  • Light—solar cell
  • Wind—windmill

Three devices for using electrical energy to generate:

  • Light—LED
  • Motion—motor
  • Sound—buzzer

Specifications

Grade 5+
For Use With (Application) Demonstrates the conversion among forms of energy
Includes Set of cables for connecting devices in various combinations, manual
Class Size 1 to 2 Lab Groups
Product Type Energy Conversion Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.