Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Equal Mass Slotted Weight Hanger
SDPSCI_ED

Catalog No. S001369 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Hanger
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S001369 Hanger
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S001369 Supplier United Scientific Supplies Supplier No. WSEQ10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

Hanger includes nine 10 g slotted masses and a 10 g hanger. Stainless steel components feature engraved values for quick identification.

Stainless steel hanging mass set includes nine removable 10 g slotted masses plus a 10 g hanger, providing a maximum total mass of 100 g. The hanger is matched to the mass of each slotted weight for simple, incremental load changes, while engraved nominal mass markings support quick identification during use.

  • Ten stainless steel components
  • Total mass of 100 g
  • Nominal mass engraved on each component
  • Central hole keeps slotted masses securely seated on the hanger
  • Supports experiments requiring equal mass changes or stepwise loading

Specifications

Product Type Hanger
Material Stainless Steel
Weight (Metric) 100 g
Includes Nine removable 10 g slotted masses and 10 g hanger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.