Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Folding Magnifer, Large
SDPSCI_ED

Catalog No. S001327 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Magnification:
10x
5x
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S001327 10x
S001312 5x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Prop 65
Catalog No. S001327 Supplier United Scientific Supplies Supplier No. MPD038
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

Large folding magnifier is easy to use.

  • Large magnifier features plastic frame with double lenses, each 38 mm (1.5 in.) in diameter
  • Each lens provides 5x magnification, for total of 10x
  • Lens diameter: 38 mm

Specifications

Product Type Magnifying Glass
Diameter (Metric) Lens 38 mm
Magnification 10x
Material Glass, Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.