Learn More
United Scientific Supplies United Scientific™ Folding Magnifier 10X, Aluminum Case
SDPSCI_ED
Catalog No. S001342 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Magnification:
10x
Catalog No. Magnification
S001342 10x
1 options
Prop 65
Catalog No. S001342 Supplier United Scientific Supplies Supplier No. MPF010
Add to Cart
Add to Cart
Prop 65
Folding magnifier provides compact 10x viewing with a protected glass lens.
  • Single folding magnifier folds into a protective aluminum case
  • Features a glass 10x magnification lens, 25 mm in diameter and mounted in a plastic frame
  • Material: Glass
  • Lens diameter: 25 mm
  • Magnification: 10x
Specifications
Product Type Magnifying Glass
Diameter (Metric) Lens 25 mm
Magnification 10x
Material Plastic