Learn More
United Scientific Supplies United Scientific™ Folding Magnifier
Shop All United Scientific Supplies, Inc Products
Description
Folding magnifiers in plastic cases. Glass lenses are 25 mm in diameter with 5x magnification. Single magnifier includes one lens. Double magnifier includes two lenses for 10x magnification. Triple magnifier includes three lenses for 15x magnification.
- Folding magnifiers with 25 mm glass lenses and plastic cases
- Single lens provides 5x magnification
- Double and triple versions offer 10x and 15x magnification respectively
Specifications
Specifications
| Product Type | Magnifying Glass |
| Material | Plastic |
| Magnification | 15x |
| Color | Black |
| Diameter (Metric) Lens | 25 mm |