Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Globe, Tilt Mounted
SDPSCI_ED

Catalog No. S001393 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Diameter (Metric):
31.75 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S001393 31.75 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S001393 Supplier United Scientific Supplies Supplier No. EARTHGLB32
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

Tilt mounted globe has labeled countries capitals political boundaries and ocean currents for geography study.

  • Diameter 31.75 cm (12.5 in.)
  • Ocean currents are illustrated
  • Total height is approximately 43 mm (17 in.)
  • Curved supporting arc includes angle measurements to help identify latitude, tilt, and seasonal changes

Specifications

Diameter (Metric) 31.75 cm
Diameter (English) 12.5 in.
Product Type Model, Earth Globe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.