Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Magnetic Wands
SDPSCI_ED

Catalog No. S001382 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W):
8 x 1 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S001382 8 x 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S001382 Supplier United Scientific Supplies Supplier No. UNMGTWNDPK24
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

ABS plastic wands with ceramic magnets are sized for easy handling and supplied in assorted colors for group activities.

Easy-to-hold ABS plastic wands contain a ceramic magnet for classroom magnetic activities. Each wand measures 8 in. long by 1 in. wide and can be used to test which items are attracted to magnets. Sold as a set of six wands in assorted colors or as a set of 24.

  • Easy-to-hold ABS plastic wands
  • Ceramic magnet encased in plastic
  • Measures 8 in. long by 1 in. wide
  • Supports simple classroom magnetic activities
  • Available as a set of six assorted-color wands or a set of 24

Specifications

Material Plastic
Shape Bar
Grade Level Elementary, Intermediate - Middle School
Product Type Magnetic Wand Set
Dimensions (L x W) 8 x 1 in.
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.