Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Plane Acrylic Mirror Strip
SDPSCI_ED

Catalog No. S001314 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Thin Square Mirror
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S001314 Thin Square Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S001314 Supplier United Scientific Supplies Supplier No. MSA4X4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

Plane acrylic mirror strip supports reflection symmetry and optics activities.

  • Plane acrylic mirror strip, 4 in. x 4 in. x 3 mm thick
  • Material: Acrylic
  • Product type: Mirror Strip

Specifications

Material Acrylic
Size 4 x 4 in.
Product Type Thin Square Mirror
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.