Learn More
United Scientific Supplies United Scientific™ Polypropylene Utility Funnel
Shop All United Scientific Supplies, Inc Products
Description
Funnels offering high quality at extraordinary value.
- Available in three sizes
- These funnels have eight internal ribs to facilitate filtration
- They are made of polypropylene
Specifications
Specifications
| Capacity (Metric) | 35, 52, 65 mm |
| Material | Polypropylene |
| Autoclavable | Autoclavable |
| Sterility | Non-sterile |
| Product Type | Utility Funnel |