Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Rubber Magdeburg Hemispheres
SDPSCI_ED

Catalog No. S001310 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Activity Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S001310 Activity Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S001310 Supplier United Scientific Supplies Supplier No. MGH002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

Rubber magdeburg hemispheres demonstrate air pressure and vacuum effects.

  • Secure two-hole handles for pulling
  • Easy release knob
  • 8.5 cm diameter suction cups

Specifications

Grade Level Intermediate - Middle School, High School, Higher Ed
Class Size 1 Lab Group
Product Type Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.