Learn More
United Scientific Supplies United Scientific™ Scalpel Blade, 10/Pk.

Shop All United Scientific Supplies, Inc Products


Description
- Available in straight, curved, and angled blade shapes
- Approximate overall length of 6.4 in.
- Approximate blade length of 2 in.
- Stainless steel construction
- Compatible scalpel handles support up to 9 blade sizes
Specifications
Specifications
| Product Type | Scalpel Blade |
| Size | # 20 |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.