Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Scalpel, Fixed Blade
SDPSCI_ED

Catalog No. S001332 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (English):
6.4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S001332 6.4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001332 Supplier United Scientific Supplies Supplier No. SCLPL1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stainless steel cutting tool with fixed blade.

  • Scalpel fixed blade
  • The characteristic of each blade lends itself to different procedures
  • Size: 6.4 in. Overall; 2 in. Blade

Specifications

Blade Material Stainless Steel
Product Type Scalpel
Length (English) Blade 2 in.
Length (English) 6.4 in.
Disposable Reusable
Sterility Non-sterile
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.