Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies United Scientific™ Screw Compressor Tubing Clamp
SDP

Catalog No. 01672033 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Diameter (English):
0.75 in.
1 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
01-672-033 0.75 in.
01-672-030 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-672-033 Supplier United Scientific Supplies Supplier No. HSCOP1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc-plated clamps control flow through flexible tubing in laboratory setups. Screw pressure adjustable and available in multiple opening sizes for different tubing needs.

  • Swivel-type screw compressor clamp constructed from zinc-plated steel wire
  • Opens up to 0.75 inch
  • Material: Zinc-Plated Steel

Specifications

Autoclavable Not Autoclavable
Type Tubing Clamp
Material Zinc-plated Steel
Diameter (English) 0.75 in.
Color Silver
Jaw Type Swivel
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.