Learn More
United Scientific Supplies United Scientific™ Single and Double Slit, Set of 3
SDPSCI_ED
Catalog No. S001362 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Type:
Multispot Slide
Catalog No. Type
S001362 Multispot Slide
1 options
Catalog No. S001362 Supplier United Scientific Supplies Supplier No. DBLESLIT3
Add to Cart
Add to Cart
Single and double slit set of 3 helps demonstrate light diffraction and interference patterns.
  • Single and double slit diffraction demonstrate the wave behavior of light
  • A single slit diffraction creates varying bright central line
  • Double slit diffraction produces an interference pattern with regularly spaced bright and dark lines
Specifications
Length (Metric) 50 mm
Width (Metric) 50 mm
Frosted Not Frosted
Type Multispot Slide
Color Gray