Learn More
United Scientific Supplies United Scientific™ Spark Timer Roll
SDPSCI_ED
Catalog No. S001317 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Tape Dispenser (Single-roll)
Catalog No. Product Type
S001317 Tape Dispenser (Single-roll)
1 options
Catalog No. S001317 Supplier United Scientific Supplies Supplier No. SPARKTAPE
  • $5.20
    $4.65 / Each of 1
    Save $0.55
    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Spark timer tape help record positions at regular intervals for distance and time analysis.
Spark Timer is used for measuring velocity and acceleration of falling or moving objects. When tape is pulled through the timer, a spark burns tiny dots in the tape at precise intervals. The faster the tape is pulled, the greater the distance between the dots, allowing students to calculate acceleration.
  • Measures velocity and acceleration of falling or moving objects
  • Creates spark marks on tape at precise intervals
  • Dot spacing helps students calculate acceleration
  • Includes spark tape, carbon disks, weight, and clamp
  • Includes activity guide
Specifications
Product Type Tape Dispenser (Single-roll)
Color White
Holds Tape
Sterility Non-sterile