Learn More
United Scientific Supplies United Scientific™ Streak Plates

Shop All United Scientific Supplies, Inc Products


Description
- Pack of 12 unglazed black streak plates, each 1 x 2 in. size
- Designed to test hardness of rocks and minerals
Specifications
Specifications
| Quantity | 12/Pk. |
| For Use With (Application) | Test hardness of rocks and minerals |
| Product Type | Streak Plate |
| Color | Black |
| Grade Level | Intermediate - Middle School, High School |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.