Learn More
United Scientific Supplies Supplied Friction on an Inclined Plane Set
SDPSCI_ED
Catalog No. S05814 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Friction on an inclined plane; Wooden inclined board: 60 x 10 cm (L x H)Inclined Plane Set FSE
Catalog No.
S05814 Friction on an inclined plane; Wooden inclined board: 60 x 10 cm (L x H)Inclined Plane Set FSE
1 options
Catalog No. S05814 Supplier United Scientific Supplies Supplier No. INPW02
Only null left
Add to Cart
Add to Cart
Includes
Inclined plane with a pulley and protractor, a Hall's carriage, a hanging pan to hold the desired weight, two wooden boards of different sizes and a metal plate
Used to demonstrate the resolution of forces on an inclined plane, uniform and accelerated motions, kinetic and potential energies and the friction between two surfaces.
  • The wooden inclined board can be set at any angle between 0° and 45°
  • Two scales on the protractor allow angle measurements to be made directly or by trigonometry
  • Students can determine the coefficients of static and sliding friction and verify the normal and frictional components of the force exerted by the plane
  • Aluminum wheel and cone bearings on the Hall's carriage help minimize friction on the inclined plane
  • Instruction manual included
Order Info
Wooden inclined board length: 23.6 in. (60cm), width: 4 in. (10cm)
Specifications
Product Type Friction on an Inclined Plane
For Use With (Application) Demonstrate the resolution of forces on an inclined plane, uniform and accelerated motions, kinetic and potential energies and the friction between two surfaces
Includes Inclined plane with a pulley and protractor, a hall's carriage, a hanging pan to hold the desired weight, two Wooden boards of different sizes and a metal plate