Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Building and Designing Batteries NextGEN STEM Kit
SDPSCI_ED

Catalog No. s13123 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S13123 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13123 Supplier United Scientific™ Supplier No. AISBAKIT
Only null left
Add to Cart
Add to Cart

Building and Designing Batteries NextGEN STEM Kit features hands-on learning materials and interactive digital content that can be accessed on computer, tablet or mobile device.

  • Enough materials for 40 students working in groups of 4.

Specifications

Grade 5+
For Use With (Application) Building and Designing Batteries
Includes Enough materials for 40 students working in groups of 4, and a DVD with PDF Teacher and Student Guides
Class Size 40 Students
Product Type NextGEN STEM Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.