Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Foucault's Pendulum
SDPSCI_ED

Catalog No. S06990 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06990 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06990 Supplier United Scientific™ Supplier No. FCPN01

missing translation for 'validMainframeMsg'

This Foucault's Pendulum is a display version of the Foucault 1851 experiment to demonstrate the Earth's rotation.

  • 57 in. high unit.
  • Contains a maintained pendulum.
  • Precession rate can be used to calculate latitude or time.

Specifications

Grade 5+
Format Kit
Product Type Foucault's Pendulum
Height (English) 57 in.
Height (Metric) 144.8 cm
Languages English
For Use With (Application) Used to demonstrate the Earth's rotation and can also be used to calculate latitude or time.
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.