Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Franck-Hertz Apparatus
SDPSCI_ED

Catalog No. S06987 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06987 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06987 Supplier United Scientific™ Supplier No. HFHA002
Add to Cart
Add to Cart

The Franck-Hertz Apparatus is a set of equipment used for observing the excitation transitions of electrons in Argon.

  • Compact unit.
  • Oscilloscope required.

Specifications

Grade 5+
Format Class Set
Product Type Franck Hertz Apparatus
Includes Argon tube, power supplies and amplifier in one box
Languages English
For Use With (Application) For observing the excitation transitions of electrons in Argon
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.