Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Heat Conductometer
SDPSCI_ED

Catalog No. S12874 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S12874 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12874 Supplier United Scientific™ Supplier No. CNDM01
Add to Cart
Add to Cart

Heat Conductometer demonstrates differences in heat conductivity of aluminum, brass, steel, nickel, and copper.

  • Used to teach concepts of transmission of heat, conductors and insulators, conductivity of a material, and the relation between conductivity and atomic structure.
  • Features five radial spokes attached to brass hub.
  • Each rod is marked with metal name on the hub for indentification.
  • Notches in each rod to contain wax pellet.
  • Unit comes assembled with supply of wax and activity guide.
  • Overall dimensions: 38 x 20 x 2.5 cm (14 x 7 x 1 in.)

Specifications

Grade 5+
Includes Wooden handle, Support rod, Copper rod (C) 4 Nickel rod (N), Steel rod, Brass rod (B), Aluminum rod (A), Brass Hub, Activity guide
Dimensions (L x W x H) 38 x 20 x 2.5 cm (14 x 7 x 1 in.)
Class Size 1 Lab Group
Product Type Heat Conductometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.