Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Thermal Emission Cans - Set of Two
SDPSCI_ED

Catalog No. S07020 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S07020 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07020 Supplier United Scientific™ Supplier No. RDCN02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrates the difference in cooling rates of hot water.

  • Two identical sized metal cans with screw cap (one smooth metal and other painted dull black)
  • Different surfaces demonstrate the difference in cooling rates of hot water inside the cans attributed to thermal radiation emitted by two cans
  • Includes activity guide

Specifications

Product Type Thermal Emission Can Set
Description Smooth Metal, Dull Black
Capacity (Metric) 1 L
Color Black, Silver
For Use With (Application) To demonstrate the difference in cooling rates of hot water
Material Metal
Includes Activity Guide, Screw Caps
Quantity 2 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.