Learn More
United Scientific Supplies United Scientific™ Sympathetic Resonance Box Set 440 Hz

Shop All United Scientific Supplies, Inc Products


Description
- Demonstrate resonance and interference
- Adjustable frequency clip enables beat frequency interference studies
- for physics experiments covering wave properties and acoustics
- Includes activation hammer and suspended ping pong ball for visual demonstrations
- Includes activity guide
Specifications
Specifications
| Grade Level | Intermediate - Middle School, High School, Higher Ed |
| Grade | 5 to 12 |
| Class Size | 1 Lab Group |
| Product Type | Resonance Demonstrator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.