Learn More
United Scientific Supplies United Scientific™ UniStore™ Reagent Bottle, Narrow Mouth
SDPSCI_ED
Catalog No. S001371 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
90 mL
Catalog No. Capacity (Metric)
S001371 90 mL
1 options
Catalog No. S001371 Supplier United Scientific Supplies Supplier No. BNM090
Add to Cart
Add to Cart
Narrow mouth HDPE bottle includes a lined polypropylene cap for controlled dispensing storage and classroom laboratory use.
Reagent bottle is equipped with a lined cap. This narrow-mouth bottle has a capacity of 90 mL and a lined polypropylene lid.
  • Constructed of high-density polyethylene
  • Translucent, pliable bottle with excellent strength
  • Leakproof design meets TSA 3-1-1 guidelines for domestic and international travel
  • Lined polypropylene lid
  • Certificates of Analysis available upon request
Specifications
Capacity (Metric) 90 mL
Color Translucent
Material HDPE
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Autoclavable Not Autoclavable
Shape Cylindrical
Disposable Reusable
Product Type Reagent Bottle