Learn More
Unity Lab Services™ Pipette Support Plans
GREENER_CHOICE
Catalog No. 12102106
Change view
Click to view available options
Calibration Points:
2 x 10 Measurements
2 x 5 Measurements
One Additional Test Volume
For Use With (Equipment):
Multi-channel Pipette
Single or Multi-channel Pipette
Single-channel Pipette
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Points For Use With (Equipment)
12-102-106 2 x 10 Measurements Multi-channel Pipette
12-102-105 2 x 10 Measurements Single-channel Pipette
12-102-103 2 x 5 Measurements Single-channel Pipette
12-102-104 2 x 5 Measurements Multi-channel Pipette
12-102-111 One Additional Test Volume Single or Multi-channel Pipette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-102-106 Supplier Unity Lab Services™ Supplier No. VFC2X10DMC
Add to Cart
Add to Cart
Fast, quality-minded service that meets your diverse needs
Whether cGLP, cGMP, critical research, academia, or any special requirements you may need, our experienced service personnel are highly trained in all pipette brands and models. We provide calibration and a detailed 15-point pipette inspection of single and multichannel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally. You will feel confident that your pipettes will have the accuracy and performance you require, when you need them.
Unity Pipette Services offers:
  • Service for any pipette brand and model
  • Reliable service options by our highly trained technicians
  • Fast and convenient prepaid pipette shipping packages
Basic – Validation-Fast Check
The Validation-Fast Check Plan combines a thorough external pipette inspection with gravimetric validation to determine if desired accuracy and precision is achieved. This plan includes a validation pass/fail report.
Ideal for customers who require:
  • An interim validation check that occurs in-between routine scheduled preventive maintenance and calibration
  • Repairs for dropped/disassembled pipettes or want to make sure pipettes are performing as desired
  • A cost-friendly service option
† If a pipette fails Validation-Fast Check, the customer can request to be contacted with a recommended service course of action (i.e. GxP Preventive Maintenance and Calibration Plan.)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multi-channel Pipette
Calibration Points 2 x 10 Measurements
Product Type Validation Fast Check, 2 x 10
Service Offering Pipette Validation
Includes Thorough external pipette inspection; Gravimetric validation to determine if desired accuracy and precision is achieved