Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Universal Plug Caps

Catalog No. 22899679
Change view
Click to view available options
Color:
Dark Blue
Green
Lavender
Light Blue
Orange
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-899-679 Orange
22-899-002 Green
22-899-000 Light Blue
22-899-001 Dark Blue
22-899-003 Lavender
22-899-004 Red
22-899-005 White
22-899-006 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-899-679 Supplier Dynamic Diagnostics Supplier No. 200011OR
Only null left
Add to Cart
Add to Cart

Eliminate the need to purchase size-specific caps

  • Fit most 10-, 12-, 13-, and 16mm tubes
  • Firm, leak-resistant design preserves sample integrity
  • Constructed of polyethylene

Specifications

Product Type Universal Plug Cap
Material Polyethylene
Color Orange
Size (English) 0.39 to 0.63 in.
Size (Metric) 10 to 16 mm
For Use With (Equipment) Fits 10, 12, 13 and 16 mm tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.