Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences™ Universal 7 Pipette Work Station, Model 337
SDP

Catalog No. 02912676 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Ice Blue
Mint Green
Pastel Rose
Vanilla Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-912-676 Mint Green
02-912-675 Ice Blue
02-912-677 Vanilla Yellow
02-912-678 Pastel Rose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-912-676 Supplier DWK Life Sciences™ Supplier No. W851356
Add to Cart
Add to Cart

Keep your workstation organized with this universal pipette stand for up to 7 pipettes. Available in a variety of colors.

  • Space-saving pipette stand holds up to seven single-channel pipettes of most brands
  • Made of sturdy polyamid material, easy to clean
  • Heavy bottom plate and anti-slip pads guarantee bench stability
  • The work station fits new and old generations of Socorex and other brand pipettes
TRUSTED_SUSTAINABILITY

Specifications

Description Model 337
Color Mint Green
Holds Pipettes (7 Single Channel)
Material Polyamide
Positions 7
Shape Round
For Use With (Equipment) Univeral
No. of Racks 1 Each
Includes Anti-slip pads
Product Type Pipette Work Station
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.