Learn More
Eppendorf™ Universal Buckets for S-4xUniversal Rotor
Catalog No. 13864431 Shop All Eppendorf North America Products
Change view
Click to view available options
Quantity:
2 Pk.
4 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-864-431 4 Pk.
13-864-430 2 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-864-431 Supplier Eppendorf™ Supplier No. 5895203000
Add to Cart
Add to Cart
Universal Buckets for S-4xUniversal Rotor are suitable swing-buckets for the S-4xUniversal Rotor, accepting a variety of adapters for tubes, flasks, and plates.
  • Suitable for S-4xUniversal Rotor
  • Accepts adapters for tubes, flasks, and plates
  • Variable places depending on adapter used
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) S-4xUniversal Rotor
Quantity 4 Pk.