Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Universal Chemiluminescent PARP Assay - Histone Coated Wells
SDP

Catalog No. 4676096K Shop All Novus Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-760-96K 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-760-96K Supplier R&D Systems™ Supplier No. 4676096K

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemiluminescent assay kit for candidate inhibitor screening and determination of IC50 values of PARP inhibitors.

HT Universal 96-well PARP Assay Kits measure the incorporation of biotinylated poly(ADP-ribose) onto histone proteins in a strip well format. This assay is ideal for the screening of PARP inhibitors and determining IC<sub>50</sub> values. Important features of the assay include: 1) chemiluminescent, non-radioactive format; 2) higher throughput 96 test size; and 3) sensitivity down to 0.01 Units of PARP per well.

Kit Contents:

  • PARP-HSA, 10 U/μL 50 μL
  • 20X PARP Buffer (2 vials) 2 x 500 μL
  • 10X PARP Cocktail (300 μlL)
  • Histone-Coated White Strip Wells (96 Wells)
  • 200 mM 3-Aminobenzamide (60 μL)
  • 10X Strep-Diluent (2 mL)
  • Strep-HRP (30 μL)
  • PeroxyGlow™ A (6 mL)
  • PeroxyGlow™ B (6 mL)
  • 10X Activated DNA (300 μL)

Specifications

Cross Reactivity Mammals
Quantity 96 Tests
Content And Storage Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems
Product Type Activity Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.