Learn More
Novus Biologicals™ Universal Dopamine ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP267270
Catalog No. NBP267270
Only null left
Add to Cart
Description
Description
The Universal Dopamine ELISA Kit (Colorimetric) measures universal Dopamine in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 - 2000 pg/mL
This kit recognizes DA in samples. No significant cross-reactivity or interference between DA and analogues was observed.
Biological Fluids, Plasma, Serum
Universal
Storage is content dependent.
18.75 pg/mL
ELISA Kit (Colorimetric)
ELISA
1 Kit
Dopamine
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.