Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Universal EZ-TFA Chemiluminescent Transcription Factor Assay

Catalog No. 70601 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
192 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-601 192 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-601 Supplier MilliporeSigma™ Supplier No. 70601

Includes

  • Streptavidin Coated Chemiluminescent Plate
  • Blocking Reagent
  • 10X Annealing Buffer
  • Chemiluminescent Detection Reagent
  • Chemiluminescent Reaction Buffer
  • TFA Buffer, 5X
  • Gt X Rb, HRP Secondary Antibody
  • Gt X Ms, HRP Secondary Antibody

Fast, sensitive method to detect specific transcription factor DNA binding activity in whole cell or nuclear extracts

  • Provided in a 96-well format
  • When the Capture Probe, a double stranded biotinylated oligonucleotide is incubated together, the active form of NFkB contained in the nuclear extract binds to its consensus sequence
  • After incubation, the extract/probe/buffer mixture is then directly transferred to the streptavidin coated plate
  • The biotinylated double stranded oligonucleotide bound by active NFkB protein is immobilized and any inactive, unbound material is washed away
  • The bound NFkB transcription factor subunits, p50 and p65, are detected with specific primary antibodies, a Rabbit anti-NFkB p50 (active form) and a Rabbit anti-NFkB p65
  • An HRP-conjugated secondary antibody is then used for detection and provides sensitive colorimetric detection that can be read in a spectrophotometric plate reader
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C:
  • Transcription Factor Assay Chemiluminescent Plate
  • Blocking Reagent
  • 10X Annealing Buffer
  • Chemiluminescent Detection Reagent
  • Chemiluminescent Rea
Detection Method Chemiluminescence
For Use With (Application) ELISA, Activity Assay
Quantity 192 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.