Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Universal Indicator, Bogens, Alcoholic, Ricca Chemical
SDP

Catalog No. 887516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
887516 500 mL Amber Glass Bottle
88754 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 887516 Supplier Ricca Chemical Company Supplier No. 887516
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Universal Indicator, Bogens
Name Note Alcoholic
CAS 67-63-0, 7732-18-5, 62625-21-2, 76-59-5, 60-11-7, 493-52-7
CAS Min % 63.0, 34.57, 0.06, 0.05, 0.04, 0.03
CAS Max % 66.0, 35.98, 0.07, 0.05, 0.04, 0.03
Grade Indicator
Packaging Amber Glass Bottle
Color Orange/Red
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.