Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Universal Indicator, pH 2 - 10, Ricca Chemical
SDP

Catalog No. 88704 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8870-4 120 mL Amber Glass Bottle
8870-32 1 L Amber Glass Bottle
R88700004C 4 L Amber Glass Bottle
R887000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 8870-4 Supplier Ricca Chemical Company Supplier No. 88704
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Universal Indicator
Name Note pH 2 - 10
CAS 64-17-5, 67-56-1, 62625-21-2, 34722-90-2, 60-11-7, 493-52-7
CAS Min % 98.0, 0.12, 0.1, 0.07, 0.05, 0.02
CAS Max % 100.0, 0.13, 0.1, 0.08, 0.05, 0.03
Grade Indicator
Packaging Amber Glass Bottle
Color Dark Brown
Quantity 120 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.